Propargyltrimethylsilane 80-90% stabilised
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Trimethyl(propargyl)silane, 3-(Trimethylsilyl) propyne, Trimethyl(prop-2-yn-1-yl)silane
366630010
366630050
ACRO366630010EA
217
EUR
ACRO366630010
ACRO366630050
Propargyltrimethylsilane 80-90% stabilised
Propargyltrimethylsilane
Formula:
C₆H₁₂Si MW: 112,25 g/mol Boiling Pt: 91…93 °C (760 mmHg) Density: 0,75 g/cm³ Flash Pt: –10 °C Storage Temperature: Freezer |
MDL Number:
MFCD00042922 CAS Number: 13361-64-3 EINECS: 236-427-6 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | 80 to 90 % |
Stabilizer | 0.1 % hydroquinone |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...