1-(Trimethylsilyl)-1-propyne ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Trimethyl-1-propynylsilane
B24875.03
B24875.06
B24875.14
B24875.03EA
28.4
EUR
B24875.03
B24875.06
B24875.14
1-(Trimethylsilyl)-1-propyne ≥98%
1-(Trimethylsilyl)-1-propyne
Purity: 98%
CAS No.: 6224-91-5
Molecular Formula: CH3C≡CSi(CH3)3
Formula Weight: 112.25
Flash Point: -3°(26°F)
Refractive Index: 1.4200
Melting Point: -69°
Boiling Point: 99-100°
Beilstein Reference: 1071311
UN No.: 1993
MDL No.: MFCD00009271
Formula:
C₆H₁₂Si MW: 112,25 g/mol Boiling Pt: 99…100 °C Melting Pt: –69 °C Density: 0,753 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009271 CAS Number: 6224-91-5 EINECS: 228-314-5 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...