1-Acetylimidazole ≥98%

Supplier: Thermo Scientific

Synonyms: 1-(1H-Imidazol-1-yl)ethanone, 1-Acetyl-1H-imidazole

L00137.06 L00137.22
L00137.06EA 0 EUR
L00137.06 L00137.22
1-Acetylimidazole ≥98%
1-Acetylimidazole

Purity: 98% CAS No.: 2466-76-4 Molecular Formula: C5H6N2O Formula Weight: 110.12 Melting Point: 98-104° Beilstein Reference: 108425

Formula: C₅H₆N₂O
MW: 110,12 g/mol
Melting Pt: 98…104 °C
MDL Number: MFCD00005287
CAS Number: 2466-76-4
EINECS: 219-577-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR