1,4-Dibromooctafluorobutane ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,4-Dibromoperfluorobutane

L16736.06 L16736.14
L16736.06EA 0 EUR
L16736.06 L16736.14
1,4-Dibromooctafluorobutane ≥98%
1,4-Dibromooctafluorobutane

Purity: 98% CAS No.: 335-48-8 Molecular Formula: C4Br2F8 Formula Weight: 359.84 Refractive Index: 1.3520 Boiling Point: 96-98° MDL No.: MFCD00153112

Formula: C₄Br₂F₈
MW: 359.84 g/mol
Boiling Pt: 96…98 °C
Density: 2.098
Storage Temperature: Ambient
MDL Number: MFCD00153112
CAS Number: 335-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR