n-Pentadecane 99%

Supplier: Thermo Fisher Scientific
Danger

129822500 129820250 129820010
ACRO129822500EA 223 EUR
ACRO129822500 ACRO129820250 ACRO129820010
n-Pentadecane 99%
n-Pentadecane
Formula: H₃C(CH₂)₁₃CH₃
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 9,9 °C
Density: 0,773 g/cm³
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00008990
CAS Number: 629-62-9
EINECS: 211-098-1
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4305 to 1.4335 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR