n-Pentadecane, Supelco®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
76509-25ML
76509-5ML
76509-25MLEA
601
EUR
76509-25ML
76509-5ML
n-Pentadecane, Supelco®
n-Pentadecane
Formula:
H₃C(CH₂)₁₃CH₃ Boiling Pt: 271 °C (1013 hPa) Melting Pt: 13…15 °C Density: 0,768 g/cm³ (20 °C) Flash Pt: 132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008990 CAS Number: 629-62-9 EINECS: 211-098-1 UN: 3295 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...