n-Triacontane, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

90270-1G 90270-250MG
90270-1GEA 199 EUR
90270-1G 90270-250MG
n-Triacontane, Supelco®
n-Triacontane
Formula: H₃C(CH₂)₂₈CH₃
Boiling Pt: 449,7 °C (1013 hPa)
Melting Pt: 65…67 °C
Density: 0,7797 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009410
CAS Number: 638-68-6
EINECS: 211-349-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=748eb4270d69d59f30cf6049c88f6e017abf84dc&vid=f60b819e90817869053b05819699c065c0bd7d76&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR