2-Methylbutyraldehyde 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
304470500
304472500
ACRO304470500EA
61
EUR
ACRO304470500
ACRO304472500
2-Methylbutyraldehyde 95%
2-Methylbutyraldehyde
Formula:
C₅H₁₀O Boiling Pt: 90 °C (1013 hPa) Melting Pt: –100 °C Density: 0,805 g/cm³ Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006984 CAS Number: 96-17-3 EINECS: 202-485-6 UN: 3371 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3860 to 1.3940 (20°C, 589 nm) |
Density | (20°C) 0.8 to 0.81 g/cm³ |
GC | ≥94 % |
Acid value | ≤10 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...