2-Chloro-2-methylpropane ≥98%

Supplier: Thermo Fisher Scientific
Danger

A13004.AK A13004.AP A13004.AU
A13004.AKEA 32.1 EUR
A13004.AK A13004.AP A13004.AU
2-Chloro-2-methylpropane ≥98%
2-Chloro-2-methylpropane

Purity: 98+% CAS No.: 507-20-0 Molecular Formula: (CH3)3CCl Formula Weight: 92.57 Flash Point: -27°(-16°F) Refractive Index: 1.3848 Odour: Mild Melting Point: -26° Boiling Point: 50-52° Beilstein Reference: 1730872 Merck Reference: 14,1562 UN No.: 1127 MDL No.: MFCD00000816

Formula: C₄H₉Cl
Boiling Pt: 50…52 °C
Melting Pt: –26 °C
Density: 0,846
Flash Pt: –27 °C (–16 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000816
CAS Number: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3,II
Merck Index: 14,1562

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR