2,3,4-Trimethylpentane ≥98%

Supplier: Thermo Fisher Scientific

L08176.06 L08176.14
L08176.06EA 72.3 EUR
L08176.06 L08176.14
2,3,4-Trimethylpentane ≥98%
2,3,4-Trimethylpentane

Purity: 98+% CAS No.: 565-75-3 Molecular Formula: C8H18 Formula Weight: 114.23 Flash Point: 5°(41°F) Refractive Index: 1.4040 Melting Point: -110° Boiling Point: 113-115° Beilstein Reference: 1696869 UN No.: 1262 MDL No.: MFCD00008924

Formula: C₈H₁₈
MDL Number: MFCD00008924
CAS Number: 565-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR