2-Methylbutane, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
iso-Pentane
8113.1000EA
84.81
EUR
8113.1000
8113.9025
2-Methylbutane, BAKER, Laboratory Reagent, J.T.Baker®
2-Methylbutane
Formula:
CH₃CH₂CH(CH₃)₂ MW: 72,15 g/mol Boiling Pt: 28 °C (1013 hPa) Melting Pt: –160 °C Density: 0,62 g/cm³ (20 °C) Flash Pt: –51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009338 CAS Number: 78-78-4 EINECS: 201-142-8 UN: 1265 ADR: 3,I |
Specification Test Results
Assay (by GC) | min. 99 % |
Boiling Point | 27.5 - 28.5 °C |
Residue after Evaporation | max. 0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...