1-Pentene 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
alpha-n-Amylene, Propylethylene
268665000
268660025
268660250
268661000
ACRO268665000EA
249
EUR
ACRO268665000
ACRO268660025
ACRO268660250
ACRO268661000
1-Pentene 97%
1-Pentene
Formula:
C₅H₁₀ Boiling Pt: 30 °C (760 mmHg) Melting Pt: –165 °C Density: 0,64 g/cm³ Flash Pt: –28 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003567 CAS Number: 109-67-1 EINECS: 203-694-5 UN: 1108 ADR: 3,I Merck Index: 12,07262 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3704 to 1.3724 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...