trans-2-Octene ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: (E)-2-Octene

L12606.06 L12606.14
L12606.06EA 90.9 EUR
L12606.06 L12606.14
trans-2-Octene ≥97%
trans-2-Octene

Purity: 97% CAS No.: 13389-42-9 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: 14°(57°F) Refractive Index: 1.4130 Melting Point: -89 to -86° Boiling Point: 122-123° Beilstein Reference: 1719497 UN No.: 3295 MDL No.: MFCD00009532

Formula: C₈H₁₆
MW: 112,22 g/mol
Boiling Pt: 122…123 °C
Melting Pt: –89 to –86 °C
Density: 0,718
Flash Pt: 14 °C (57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009532
CAS Number: 13389-42-9
EINECS: 236-463-2
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR