1-Heptyne ≥99%
Supplier: Thermo Scientific
|
Danger
|
A11130.06
A11130.14
A11130.22
A11130.06EA
30.4
EUR
A11130.06
A11130.14
A11130.22
1-Heptyne ≥99%
1-Heptyne
Purity: 99%
CAS No.: 628-71-7
Molecular Formula: CH3(CH2)4C≡CH
Formula Weight: 96.17
Flash Point: -10°(14°F)
Refractive Index: 1.4095
Odour: Unpleasant
Melting Point: -81°
Boiling Point: 98-100°
Beilstein Reference: 1733518
UN No.: 3295
MDL No.: MFCD00009529
Formula:
C₇H₁₂ MW: 96,17 g/mol Boiling Pt: 98…100 °C Melting Pt: –81 °C Density: 0,736 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009529 CAS Number: 628-71-7 EINECS: 211-051-5 UN: 3295 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...