3-Hexyne 99%

Supplier: Thermo Scientific
Danger

308120250 308120050
ACRO308120250EA 125 EUR
ACRO308120250 ACRO308120050
3-Hexyne 99%
3-Hexyne
Formula: C₆H₁₀
MW: 82,15 g/mol
Boiling Pt: 81…82 °C
Melting Pt: –61,5 °C
Density: 0,72 g/cm³
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009381
CAS Number: 928-49-4
EINECS: 213-173-4
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR