9-Anthrealdehyde 99%

Supplier: Thermo Fisher Scientific

153930250 153931000 153935000
ACRO153930250EA 75 EUR
ACRO153930250 ACRO153931000 ACRO153935000
9-Anthrealdehyde 99%
9-Anthrealdehyde
Formula: C₁₅H₁₀O
Boiling Pt: 369 °C (1013 hPa)
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001254
CAS Number: 642-31-9
EINECS: 211-383-0

Order Now

Specification Test Results

Appearance Yellow to yellow-green to yellow-brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 103 to 107 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR