9-Anthrealdehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
9-Anthracenecarbaldehyde, 9-Anthracenecarboxaldehyde, 9-Anthraldehyde
A11258.14
A11258.22
A11258.36
A11258.14EA
63
EUR
A11258.14
A11258.22
A11258.36
9-Anthrealdehyde ≥98%
9-Anthrealdehyde
Purity: 98%
CAS No.: 642-31-9
Molecular Formula: C15H10O
Formula Weight: 206.24
Melting Point: 102-106°
Beilstein Reference: 639167
Formula:
C₁₅H₁₀O MW: 206.24 g/mol Boiling Pt: 369 Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001254 CAS Number: 642-31-9 EINECS: 211-383-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...