9,10-Dimethylanthracene 97%

Supplier: Thermo Fisher Scientific

115970010 115972500
ACRO115970010EA 138 EUR
ACRO115970010 ACRO115972500
9,10-Dimethylanthracene 97%
9,10-Dimethylanthracene
Formula: C₁₆H₁₄
MW: 206.29 g/mol
Boiling Pt: 371
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00001262
CAS Number: 781-43-1
EINECS: 212-308-4

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 180 to 185 °C
GC ≥96.0 %
Acidity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR