4-Bromobenzoic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: p-Bromobenzoic Acid

A12518.14 A12518.22 A12518.36
A12518.14EA 46.4 EUR
A12518.14 A12518.22 A12518.36
4-Bromobenzoic acid ≥98%
4-Bromobenzoic acid

Purity: 98+% CAS No.: 586-76-5 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Odour: Odourless Melting Point: 253-258° Beilstein Reference: 1906923 Merck Reference: 14,1408 MDL No.: MFCD00002529

Formula: C₇H₅BrO₂
MW: 201,02 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 253…258 °C
Density: 1,894
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7
Merck Index: 14,1408

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR