5-Bromoanthranilic acid 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Amino-5-bromobenzoic acid
A11915.22
A11915.06
A11915.14
A11915.22EA
502
EUR
A11915.22
A11915.06
A11915.14
5-Bromoanthranilic acid 98%
5-Bromoanthranilic acid
Purity: 98%
CAS No.: 5794-88-7
Molecular Formula: C7H6BrNO2
Formula Weight: 216.03
Melting Point: ca 215° dec.
Beilstein Reference: 639028
Merck Reference: 14,1405
MDL No.: MFCD00007823
Formula:
C₇H₆BrNO₂ MW: 216,03 g/mol Boiling Pt: 342 °C (1013 hPa) Melting Pt: ∼215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007823 CAS Number: 5794-88-7 EINECS: 227-338-3 UN: 2811 ADR: 6.1,III Merck Index: 14,1405 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...