2-Bromo-5-methoxybenzoic acid ≥98%

Supplier: Thermo Scientific
Warning

A12261.18 A12261.09 A12261.30
A12261.18EA 178 EUR
A12261.18 A12261.09 A12261.30
2-Bromo-5-methoxybenzoic acid ≥98%
2-Bromo-5-methoxybenzoic acid

Purity: 98+% CAS No.: 22921-68-2 Molecular Formula: C8H7BrO3 Formula Weight: 231.05 Odour: Odourless Melting Point: 156-160° Beilstein Reference: 639763 MDL No.: MFCD00020214

Formula: C₈H₇BrO₃
MW: 231,05 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00020214
CAS Number: 22921-68-2
EINECS: 245-329-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR