1-Bromo-2-chlorobenzene ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Bromochlorobenzene
A11782.0B
A11782.18
A11782.30
A11782.0BEA
566
EUR
A11782.0B
A11782.18
A11782.30
1-Bromo-2-chlorobenzene ≥98%
1-Bromo-2-chlorobenzene
Purity: 98+%
CAS No.: 694-80-4
Molecular Formula: C6H4BrCl
Formula Weight: 191.46
Flash Point: 79°(174°F)
Refractive Index: 1.5815
Melting Point: -13°
Boiling Point: 203-205°
Beilstein Reference: 1855303
MDL No.: MFCD00000532
Formula:
C₆H₄BrCl MW: 191,45 g/mol Boiling Pt: 203…205 °C Melting Pt: –13 °C Density: 1,649 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000532 CAS Number: 694-80-4 EINECS: 211-775-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...