4-Bromo-2-chlorobenzoic acid ≥98%

Supplier: Thermo Scientific
Warning

A11527.03
A11527.03EA 22.8 EUR
A11527.03
4-Bromo-2-chlorobenzoic acid ≥98%
4-Bromo-2-chlorobenzoic acid

Purity: 98+% CAS No.: 59748-90-2 Molecular Formula: C7H4BrClO2 Formula Weight: 235.47 Melting Point: 172-174° MDL No.: MFCD00040903

Formula: C₇H₄BrClO₂
MW: 235,46 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00040903
CAS Number: 59748-90-2
EINECS: 261-911-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR