Benzophenone hydrazone 98+%
Supplier: Thermo Scientific
105571000
105570050
105575000
ACRO105571000EA
45.7
EUR
ACRO105571000
ACRO105570050
ACRO105575000
Benzophenone hydrazone 98+%
Benzophenone hydrazone
Formula:
C₁₃H₁₂N₂ MW: 196,25 g/mol Boiling Pt: 225…230 °C (55 mmHg) Melting Pt: 95…99 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007624 CAS Number: 5350-57-2 EINECS: 226-321-8 ADR: 6.1,III |
Specification Test Results
Appearance | White to yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 95 to 99 °C |
Water | ≤0.5 % |
GC | ≥98.0 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...