Ethyl oxamate 99%

Supplier: Thermo Scientific

Synonyms: Oxamic Acid Ethyl Ester, Ethyl 2-amino-2-oxoacetate

118790250 118795000
ACRO118790250EA 27.5 EUR
ACRO118790250 ACRO118795000
Ethyl oxamate 99%
Ethyl oxamate
Formula: C₄H₇NO₃
MW: 117,1 g/mol
Melting Pt: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008005
CAS Number: 617-36-7
EINECS: 210-512-8

Order Now

Specification Test Results

Appearance White Crystalline powder, crystals and/or chunks
Infrared spectrum Conforms
Melting point 114 to 119 °C
Assay ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR