Oxamic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Amino-2-oxoacetic acid
129620100
129620250
129621000
ACRO129620100EA
50.2
EUR
ACRO129620100
ACRO129620250
ACRO129621000
Oxamic acid 98%
Oxamic acid
Formula:
C₂H₃NO₃ MW: 89,05 g/mol Melting Pt: 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008006 CAS Number: 471-47-6 EINECS: 207-443-0 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Titration RCOCl | ≥96.0 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...