Oxamide 98%
Supplier: Thermo Scientific
|
Warning
|
129651000
129655000
ACRO129651000EA
41.9
EUR
ACRO129651000
ACRO129655000
Oxamide 98%
Oxamide
Formula:
C₂H₄N₂O₂ MW: 88,07 g/mol Melting Pt: 491 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008007 CAS Number: 471-46-5 EINECS: 207-442-5 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration excess NaOH | ≥97.5 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...