Diphenyl sulphone 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Phenyl sulphone, sulphonyldibenzene
130960010
130960050
130962500
ACRO130960010EA
143
EUR
ACRO130960010
ACRO130960050
ACRO130962500
Diphenyl sulphone 97%
Diphenyl sulphone
Formula:
C₁₂H₁₀O₂S MW: 218,28 g/mol Melting Pt: 125…129 °C |
MDL Number:
MFCD00007548 CAS Number: 127-63-9 EINECS: 204-853-1 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 125 to 129 °C |
GC | ≥96.0 % |
Stabilizer | Approx. 50 ppm BHT |
Peroxide value | ≤50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...