2-Vinylnaphthalene 97% stabilised
Supplier: Thermo Scientific
140900050
140900250
ACRO140900050EA
90.9
EUR
ACRO140900050
ACRO140900250
2-Vinylnaphthalene 97% stabilised
2-Vinylnaphthalene
Formula:
C₁₂H₁₀ MW: 154,21 g/mol Boiling Pt: 135 °C (18 mmHg) Melting Pt: 63…67 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004125 CAS Number: 827-54-3 EINECS: 212-573-6 |
Specification Test Results
Appearance | White to yellow to beige Powder or crystals or flakes and/or chunks |
Infrared spectrum | Conforms |
Melting point | 63 to 68 °C |
GC | ≥96.0 % |
Stabilizer | ≤0.5 % p-tert-Butylcatechol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...