6-Methylpyridin-3-ol 99%

Supplier: Thermo Scientific

Synonyms: 2-Methylpyridin-5-ol, 3-Hydroxy-6-methylpyridine, 5-Hydroxy-2-methylpyridine

148110250 148111000
ACRO148110250EA 77.8 EUR
ACRO148110250 ACRO148111000
6-Methylpyridin-3-ol 99%
6-Methylpyridin-3-ol
Formula: C₆H₇NO
MW: 109,13 g/mol
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00006339
CAS Number: 1121-78-4
EINECS: 214-337-8

Order Now

Specification Test Results

Appearance Beige to tan Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 171 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR