Nitrosium tetrafluoroborate 97%

Supplier: Thermo Scientific
Danger

Synonyms: Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate

174390250 174390050 174391000
ACRO174390250EA 254 EUR
ACRO174390250 ACRO174390050 ACRO174391000
Nitrosium tetrafluoroborate 97%
Nitrosium tetrafluoroborate
Formula: BF₄NO
MW: 116,81 g/mol
Melting Pt: 250 °C (subl.)
Density: 2,185 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011433
CAS Number: 14635-75-7
EINECS: 238-679-2
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Assay ≥96.0 % (ex F)
Melting point 75 to 83 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR