Triisopropanolamine 98%

Supplier: Thermo Scientific
Warning

245760010 245760100 245761000
ACRO245760010EA 42.4 EUR
ACRO245760010 ACRO245760100 ACRO245761000
Triisopropanolamine 98%
Triisopropanolamine
Formula: C₉H₂₁NO₃
Boiling Pt: 190 °C (23 mmHg)
Melting Pt: 48…52 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004533
CAS Number: 122-20-3
EINECS: 204-528-4

Order Now

Specification Test Results

Appearance White to light yellow Low melting crystalline solid
Infrared spectrum Conforms
Titration with HCl ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR