Titanyl(IV) acetylacetonate
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Titanium(IV) oxide acetylacetonate, Titanium(IV) oxyacetylacetonate, Titanyl Acetylacetonate, Titanium (IV) oxide bis(acetylacetonate), Oxobis(2,4-pentanedionato-O,O')titanium (IV), Titanium (IV) oxide bis(2,4-pentanedionate)
77130.09
77130.18
77130.30
77130.09EA
116
EUR
77130.09
77130.18
77130.30
Titanyl(IV) acetylacetonate
Titanyl (IV) acetylacetonate
CAS No.: 14024-64-7
Molecular Formula: C10H14O5Ti
Formula Weight: 262.12
Physical Form: Powder
Melting Point: 184° dec.
MDL No.: MFCD00013505
Formula:
C₁₀H₁₄O₅Ti MW: 262,1 g/mol Melting Pt: 184 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013505 CAS Number: 14024-64-7 EINECS: 237-861-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...