4-Fluorophenyl methyl sulphone ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Fluoro-4-(methylsulphonyl)benzene, 1-Fluor-4-(methylsulphonyl)benzol

A12863.06 A12863.14
A12863.06EA 66.5 EUR
A12863.06 A12863.14
4-Fluorophenyl methyl sulphone ≥98%
4-Fluorophenyl methyl sulfone

Purity: 98% CAS No.: 455-15-2 Molecular Formula: C7H7FO2S Formula Weight: 174.19 Melting Point: 76-81° Beilstein Reference: 2045674

Formula: C₇H₇FO₂S
MW: 174,2 g/mol
Melting Pt: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00039753
CAS Number: 455-15-2
EINECS: 207-237-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR