Tetramethylene sulphoxide ≥97%
Supplier: Thermo Scientific
Synonyms:
Tetrahydrothiophene 1-oxide
A17502.09
A17502.18
A17502.09EA
48.7
EUR
A17502.09
A17502.18
Tetramethylene sulphoxide ≥97%
Tetramethylene sulphoxide
Purity: 97%
CAS No.: 1600-44-8
Molecular Formula: C4H8OS
Formula Weight: 104.17
Flash Point: 112°(233°F)
Refractive Index: 1.5200
Boiling Point: 235-237°
Beilstein Reference: 105274
MDL No.: MFCD00005477
Formula:
C₄H₈OS MW: 104,17 g/mol Boiling Pt: 235…237 °C Density: 1,173 Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005477 CAS Number: 1600-44-8 EINECS: 216-493-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...