Cyclopropyl-4-methoxyphenyl ketone ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Cyclopropyl(4-methoxyphenyl)methanone

B20725.06 B20725.14
B20725.06EA 33.7 EUR
B20725.06 B20725.14
Cyclopropyl-4-methoxyphenyl ketone ≥98%
Cyclopropyl 4-methoxyphenyl ketone

Purity: 98% CAS No.: 7152-03-6 Molecular Formula: C11H12O2 Formula Weight: 176.22 Flash Point: 110°(230°F) Melting Point: 40-44° Boiling Point: 158-160°/4.5mm Beilstein Reference: 2046539 MDL No.: MFCD00001296

Formula: C₁₁H₁₂O₂
MW: 176,22 g/mol
Boiling Pt: 158…160 °C (4,5 mmHg)
Melting Pt: 40…44 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001296
CAS Number: 7152-03-6
EINECS: 230-487-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR