2-Chloroethyl phenyl sulphone ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
B21690.06
B21690.14
B21690.22
B21690.06EA
50.2
EUR
B21690.06
B21690.14
B21690.22
2-Chloroethyl phenyl sulphone ≥98%
2-Chloroethyl phenyl sulphone
Purity: 98%
CAS No.: 938-09-0
Molecular Formula: C8H9ClO2S
Formula Weight: 204.68
Melting Point: 52-55°
Boiling Point: 168-174°/7mm
UN No.: 1759
MDL No.: MFCD00025047
Formula:
C₈H₉ClO₂S Boiling Pt: 168…174 °C (7 mmHg) Melting Pt: 52…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025047 CAS Number: 938-09-0 EINECS: 213-337-5 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...