Methyl-4-bromo-2-methoxybenzoate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 4-bromo-2-methoxybenzoate, 4-Bromo-2-methoxybenzoic acid methyl ester

H27773.09 H27773.03
H27773.09EA 153 EUR
H27773.09 H27773.03
Methyl-4-bromo-2-methoxybenzoate ≥98%
Methyl 4-bromo-2-methoxybenzoate

Purity: 98% CAS No.: 139102-34-4 Molecular Formula: C9H9BrO3 Formula Weight: 245.08 Flash Point: 110°(230°F) Melting Point: 32-34° Boiling Point: 100-108°/0.1mm MDL No.: MFCD06797867

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Boiling Pt: 100…108 °C (0.1 mmHg)
Melting Pt: 32…34 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD06797867
CAS Number: 139102-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR