5-Methylpyridin-3-ol ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

H32658.MD
H32658.MDEA 94.3 EUR
H32658.MD
5-Methylpyridin-3-ol ≥97%
5-Methylpyridin-3-ol

Purity: 97% CAS No.: 42732-49-0 Molecular Formula: C6H7NO Formula Weight: 109.13 Melting Point: 136-140° MDL No.: MFCD00661297

Formula: C₆H₇NO
MW: 109,13 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00661297
CAS Number: 42732-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR