(±)-3-(Methylamino)-1,2-propanediol 99%

Supplier: Thermo Scientific
Danger

Synonyms: 3-(Methylamino)propane-1,2-diol, (±)-3-(Methylamino)-1,2-dihydroxy propane

L12653.14 L12653.22
L12653.14EA 50 EUR
L12653.14 L12653.22
(±)-3-(Methylamino)-1,2-propanediol 99%
(±)-3-(Methylamino)-1,2-propanediol

Purity: 99% CAS No.: 40137-22-2 Molecular Formula: C4H11NO2 Formula Weight: 105.14 Flash Point: 152°(305°F) Refractive Index: 1.4745 Boiling Point: 239-241° Beilstein Reference: 1733341 UN No.: 2735 MDL No.: MFCD00082732

Formula: C₄H₁₁NO₂
MW: 105,14 g/mol
Boiling Pt: 239…241 °C
Density: 1,093
Flash Pt: 152 °C (305 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082732
CAS Number: 40137-22-2
EINECS: 254-809-0
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR