2,6-Toluenediisocyanate ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2,6-TDI, 2-Methyl-m-phenylene diisocyanate, Tolylene 2,6-diisocyanate

L12745.03 L12745.06
L12745.03EA 80.3 EUR
L12745.03 L12745.06
2,6-Toluenediisocyanate ≥97%
2,6-Toluenediisocyanate

Purity: 97% CAS No.: 91-08-7 Molecular Formula: C9H6N2O2 Formula Weight: 174.16 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5708 Odour: Pungent Melting Point: 13° Boiling Point: 129-133°/18mm Beilstein Reference: 2211546 UN No.: 2078 MDL No.: MFCD00002010

Formula: C₉H₆N₂O₂
MW: 174,16 g/mol
Boiling Pt: 129…133 °C (18 mmHg)
Melting Pt: 13 °C
Density: 1,226
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00002010
CAS Number: 91-08-7
EINECS: 202-039-0
UN: 2078
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR