6-Chloro-m-anisidine hydrochloride ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Chloro-5-methoxyanilinium chloride, 3-Amino-4-chloroanisole hydrochloride, 2-Chloro-5-methoxyaniline hydrochloride, 6-Chloro-3-anisidine hydrochloride

L13490.06 L13490.14
L13490.06EA 64.4 EUR
L13490.06 L13490.14
6-Chloro-m-anisidine hydrochloride ≥99%
6-Chloro-m-anisidine hydrochloride

Purity: 99% CAS No.: 85006-21-9 Molecular Formula: C7H8ClNO.HCl Formula Weight: 194.06 Melting Point: ca 207° dec. Beilstein Reference: 3699386 UN No.: 2811 MDL No.: MFCD00012962

Formula: C₇H₉Cl₂NO
MW: 194,06 g/mol
Melting Pt: ∼ 207 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012962
CAS Number: 85006-21-9
EINECS: 285-050-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR