2-Bromobenzaldehyde diethyl acetal ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
1-Bromo-2-(diethoxymethyl)benzene, 2-Bromo-α,α-diethoxytoluene
L15513.06
L15513.14
L15513.22
L15513.06EA
0
EUR
L15513.06
L15513.14
L15513.22
2-Bromobenzaldehyde diethyl acetal ≥98%
2-Bromobenzaldehyde diethyl acetal
Purity: 98%
CAS No.: 35822-58-3
Molecular Formula: C11H15BrO2
Formula Weight: 259.14
Flash Point: 113°(235°F)
Refractive Index: 1.5150
Boiling Point: 95°/1mm
Beilstein Reference: 5935613
MDL No.: MFCD01075696
Formula:
C₁₁H₁₅BrO₂ Boiling Pt: 95 °C (1 mmHg) Density: 1,285 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01075696 CAS Number: 35822-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...