3-Bromo-4-fluorobenzaldehyde 99%

Supplier: Thermo Fisher Scientific
Warning

293930050 293930250
ACRO293930050EA 32.2 EUR
ACRO293930050 ACRO293930250
3-Bromo-4-fluorobenzaldehyde 99%
3-Bromo-4-fluorobenzaldehyde
Formula: C₇H₄BrFO
Boiling Pt: 138…139 °C (2,5 mmHg)
Melting Pt: 28…33 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042186
CAS Number: 77771-02-9
EINECS: 278-764-1

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 28 to 33 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR