Butyl acrylate stabilised, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
n-Butyl acrylate, Acrylic acid butyl ester
Stabilisation:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200
8.00832.0100
8.00832.0500
8.00832.2500
8.00832.0100EA
25.7
EUR
8.00832.0100
8.00832.0500
8.00832.2500
Butyl acrylate stabilised, Sigma-Aldrich®
Butyl acrylate
Formula:
C₇H₁₂O₂ MW: 128,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008809 CAS Number: 141-32-2 EINECS: 205-480-7 UN: 2348 ADR: 3,III |
Specification Test Results
Solubility | 2 g/l (20 °C) |
Melting Point | -65 °C |
Molar Mass | 128.17 g/mol |
Boiling Point | 147 - 148 °C (1013 hPa) |
Vapor Pressure | 4.3 hPa (20 °C) |
Flash Point | 36.5 °C |
Refractive Index | 1.419 (20 °C, 589 nm) |
Explosion Limit | 1.1 - 7.8 %(V) |
Density | 0.90 g/cm3 (20 °C) |
Ignition Point | 267 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...