Methyl tricosanoate, Supelco®

Supplier: Merck

Synonyms: Tricosanoic acid methyl ester

M-Clarity™ quality level = MQ100

91478-1G 91478-250MG
91478-1GEA 174 EUR
91478-1G 91478-250MG
Methyl tricosanoate, Supelco®
Methyl tricosanoate
Formula: C₂₄H₄₈O₂
MW: 368,64 g/mol
MDL Number: MFCD00009349
CAS Number: 2433-97-8
EINECS: 219-420-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed46e790e4b7382def4a0e22cd9279064f2f6b20&vid=ca4db1fc8b0caecb5750e9ade94de37422f99a1d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR