1-Bromo-4-iodobenzene 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Bromo-1-iodobenzene
218380250
218381000
ACRO218380250EA
124
EUR
ACRO218380250
ACRO218381000
1-Bromo-4-iodobenzene 98%
1-Bromo-4-iodobenzene
Formula:
C₆H₄BrI Boiling Pt: 120…122 °C (14 mmHg) Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001051 CAS Number: 589-87-7 EINECS: 209-662-7 |
Specification Test Results
Appearance | White to brown Crystalline powder or crystals or needles |
Infrared spectrum | Conforms |
Melting point | 89 to 93 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...