2-Bromoaniline ≥98%

Supplier: Thermo Scientific
Danger

A11788.14 A11788.22 A11788.36
A11788.14EA 46.9 EUR
A11788.14 A11788.22 A11788.36
2-Bromoaniline ≥98%
2-Bromoaniline

Purity: 98% CAS No.: 615-36-1 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 110°(230°F) Odour: Characteristic Boiling Point: 228-230° UN No.: 2811 MDL No.: MFCD00007632

Formula: C₆H₆BrN
MW: 172,02 g/mol
Boiling Pt: 228…230 °C
Melting Pt: 27…31 °C
Density: 1,578
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007632
CAS Number: 615-36-1
EINECS: 210-421-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR