3-Bromoaniline 98%

Supplier: Thermo Fisher Scientific
Warning

177210050 177210250 177211000
ACRO177210050EA 30.8 EUR
ACRO177210050 ACRO177210250 ACRO177211000
3-Bromoaniline 98%
3-Bromoaniline
Formula: C₆H₆BrN
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 16,8 °C
Density: 1,58 g/cm³
Flash Pt: >230 °C
Storage Temperature: Ambient
MDL Number: MFCD00007757
CAS Number: 591-19-5
EINECS: 209-704-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear light yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6240 to 1.6270 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR