1-Bromo-4-nitrobenzene 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Bromonitrobenzene
173360100
173360250
173361000
ACRO173360100EA
42.7
EUR
ACRO173360100
ACRO173360250
ACRO173361000
1-Bromo-4-nitrobenzene 98%
1-Bromo-4-nitrobenzene
Formula:
C₆H₄BrNO₂ MW: 202,01 g/mol Boiling Pt: 255 °C (1013 hPa) Melting Pt: 125…127 °C Density: 1,93 g/cm³ (20 °C) Flash Pt: ≥ 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007280 CAS Number: 586-78-7 EINECS: 209-583-8 UN: 3459 ADR: 6.1,III |
Specification Test Results
Appearance | White to yellow to orange to brown Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 123 to 128 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...